The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144663.1 | NP_001138135.1 | 9 - 10 | 1189 | 74 |
NM_004063.3 | NP_004054.3 | 9 - 10 | 1188 | 74 | |
XM_011516790.1 | XP_011515092.1 | 9 - 10 | 1187 | 74 | |
GenBank mRNA | AK300638.1 | - | 1 - 2 | 67 | 74 |
AK312873.1 | - | 9 - 10 | 1189 | 74 | |
BC112013.1 | - | 9 - 10 | 1161 | 74 | |
BC113464.1 | - | 9 - 10 | 1149 | 74 | |
U07969.1 | - | 9 - 10 | 1150 | 74 | |
X83228.1 | - | 9 - 10 | 1155 | 74 |