The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018477.2 | NP_060947.1 | 1 - 2 | 181 | 111 |
XM_011536960.1 | XP_011535262.1 | 1 - 2 | 126 | 111 | |
XM_011536961.1 | XP_011535263.1 | 1 - 2 | 126 | 111 | |
XM_011536962.2 | XP_011535264.1 | 1 - 2 | 138 | 111 | |
XM_011536963.2 | XP_011535265.1 | 1 - 2 | 137 | 111 | |
GenBank mRNA | AF220190.1 | - | 1 - 2 | 158 | 111 |
AK000544.1 | - | 1 - 2 | 161 | 111 | |
AK022534.1 | - | 1 - 2 | 133 | 111 | |
AK222624.1 | - | 1 - 2 | 142 | 111 | |
AK222702.1 | - | 1 - 2 | 142 | 111 | |
AK315537.1 | - | 1 - 2 | 126 | 111 | |
BC005208.1 | - | 1 - 2 | 142 | 111 | |
BC011997.1 | - | 1 - 2 | 115 | 111 | |
BM750953.1 | - | 1 - 2 | 181 | 111 | |
BX248282.1 | - | 1 - 2 | 77 | 111 |