Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELF4
Entrez Gene ID
56853
Gene Name
CUGBP, Elav-like family member 4
Gene Aliases
BRUNOL4, CELF-4
Chromosome Location
Chr.18: 37243040 - 37566052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025087.1 NP_001020258.1 2 - 3 768 103
NM_001025088.1 NP_001020259.1 2 - 3 768 103
NM_001025089.1 NP_001020260.1 2 - 3 768 103
NM_020180.3 NP_064565.1 2 - 3 768 103
XM_005258308.3 XP_005258365.1 2 - 3 780 103
XM_011526079.2 XP_011524381.1 2 - 3 782 103
XM_011526081.2 XP_011524383.1 2 - 3 619 103
XM_011526083.2 XP_011524385.1 2 - 3 782 103
XM_011526084.2 XP_011524386.1 2 - 3 782 103
XM_011526085.2 XP_011524387.1 2 - 3 783 103
XM_011526086.2 XP_011524388.1 2 - 3 781 103
XM_011526087.2 XP_011524389.1 2 - 3 783 103
XM_011526090.2 XP_011524392.1 2 - 3 780 103
XM_011526091.2 XP_011524393.1 2 - 3 780 103
XM_011526092.2 XP_011524394.1 2 - 3 780 103
XM_017025840.1 XP_016881329.1 2 - 3 645 103
XM_017025841.1 XP_016881330.1 2 - 3 779 103
XM_017025842.1 XP_016881331.1 2 - 3 781 103
XM_017025843.1 XP_016881332.1 2 - 3 781 103
XM_017025844.1 XP_016881333.1 2 - 3 781 103
XM_017025845.1 XP_016881334.1 2 - 3 781 103
XM_017025846.1 XP_016881335.1 2 - 3 779 103
XM_017025847.1 XP_016881336.1 2 - 3 779 103
XM_017025848.1 XP_016881337.1 2 - 3 780 103
XM_017025849.1 XP_016881338.1 2 - 3 780 103
XM_017025850.1 XP_016881339.1 2 - 3 645 103
XM_017025851.1 XP_016881340.1 2 - 3 645 103
XM_017025852.1 XP_016881341.1 2 - 3 645 103
XM_017025853.1 XP_016881342.1 2 - 3 781 103
XM_017025854.1 XP_016881343.1 2 - 3 781 103
XM_017025855.1 XP_016881344.1 2 - 3 781 103
XM_017025856.1 XP_016881345.1 2 - 3 781 103
XM_017025857.1 XP_016881346.1 2 - 3 782 103
XM_017025858.1 XP_016881347.1 2 - 3 619 103
XM_017025859.1 XP_016881348.1 2 - 3 619 103
XM_017025860.1 XP_016881349.1 2 - 3 780 103
XM_017025861.1 XP_016881350.1 2 - 3 645 103
XM_017025862.1 XP_016881351.1 2 - 3 645 103
XM_017025863.1 XP_016881352.1 2 - 3 627 103
XM_017025864.1 XP_016881353.1 2 - 3 619 103
XM_017025865.1 XP_016881354.1 2 - 3 645 103
XM_017025866.1 XP_016881355.1 2 - 3 203 103
XM_017025867.1 XP_016881356.1 2 - 3 180 103
XM_017025868.1 XP_016881357.1 2 - 3 180 103
XM_017025869.1 XP_016881358.1 2 - 3 174 103
XM_017025870.1 XP_016881359.1 2 - 3 180 103
XM_017025871.1 XP_016881360.1 2 - 3 180 103
XM_017025872.1 XP_016881361.1 2 - 3 174 103
GenBank mRNA AB209774.1 - 2 - 3 562 103
AF248650.1 - 2 - 3 507 103
AF329265.1 - 2 - 3 372 103
AK074596.1 - 2 - 3 500 103
AK295009.1 - 2 - 3 382 103
BC001946.1 - 2 - 3 529 103
BC004167.2 - 2 - 3 529 103
BC045711.1 - 2 - 3 455 103

View Full Product Details