The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012434.4 | NP_036566.1 | 6 - 7 | 948 | 122 |
XM_005248710.3 | XP_005248767.1 | 6 - 7 | 970 | 122 | |
XM_005248711.2 | XP_005248768.1 | 6 - 7 | 1202 | 122 | |
XM_011535750.2 | XP_011534052.1 | 6 - 7 | 906 | 122 | |
XM_017010730.1 | XP_016866219.1 | 5 - 6 | 1114 | 122 | |
XM_017010731.1 | XP_016866220.1 | 5 - 6 | 964 | 122 | |
GenBank mRNA | AF244577.1 | - | 7 - 8 | 1205 | 122 |
AJ387747.1 | - | 6 - 7 | 1090 | 122 | |
AK294068.1 | - | 5 - 6 | 819 | 122 | |
BC020961.2 | - | 6 - 7 | 907 | 122 |