Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAVER2
Entrez Gene ID
55225
Gene Name
ribonucleoprotein, PTB binding 2
Gene Aliases
-
Chromosome Location
Chr.1: 64744979 - 64833232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018211.3 NP_060681.2 1 - 2 329 123
XM_006710738.3 XP_006710801.2 1 - 2 436 123
XM_011541706.2 XP_011540008.1 1 - 2 445 123
GenBank mRNA AB046799.1 - 1 - 2 329 123

View Full Product Details