The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005603.4 | NP_005594.1 | 18 - 19 | 2221 | 93 |
XM_006722481.3 | XP_006722544.1 | 18 - 19 | 2957 | 93 | |
XM_011526022.2 | XP_011524324.1 | 18 - 19 | 2145 | 93 | |
XM_011526023.2 | XP_011524325.1 | 17 - 18 | 2843 | 93 | |
GenBank mRNA | AF038007.1 | - | 17 - 18 | 2101 | 93 |
BC003534.1 | - | 7 - 8 | 1029 | 93 |