The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302489.1 | NP_001289418.1 | 5 - 6 | 580 | 68 |
NM_001302490.1 | NP_001289419.1 | 4 - 5 | 512 | 68 | |
NM_001610.3 | NP_001601.1 | 5 - 6 | 608 | 68 | |
GenBank mRNA | AK298439.1 | - | 5 - 6 | 583 | 68 |
AK299382.1 | - | 5 - 6 | 537 | 68 | |
AK300569.1 | - | 4 - 5 | 536 | 68 | |
AK301860.1 | - | 4 - 5 | 470 | 68 | |
AY007137.1 | - | 2 - 3 | 140 | 68 | |
BC003160.1 | - | 5 - 6 | 553 | 68 | |
BC093010.1 | - | 5 - 6 | 566 | 68 | |
X12548.1 | - | 5 - 6 | 562 | 68 |