The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302489.1 | NP_001289418.1 | 7 - 8 | 808 | 77 |
NM_001302490.1 | NP_001289419.1 | 6 - 7 | 740 | 77 | |
NM_001302491.1 | NP_001289420.1 | 5 - 6 | 647 | 77 | |
NM_001302492.1 | NP_001289421.1 | 6 - 7 | 765 | 77 | |
NM_001610.3 | NP_001601.1 | 7 - 8 | 836 | 77 | |
GenBank mRNA | AK298122.1 | - | 5 - 6 | 602 | 77 |
AK298439.1 | - | 7 - 8 | 811 | 77 | |
AK300569.1 | - | 6 - 7 | 764 | 77 | |
AK300911.1 | - | 6 - 7 | 737 | 77 | |
AK301860.1 | - | 6 - 7 | 698 | 77 | |
AK303894.1 | - | 6 - 7 | 732 | 77 | |
AK308512.1 | - | 5 - 6 | 1055 | 77 | |
AK311431.1 | - | 6 - 7 | 730 | 77 | |
AY007137.1 | - | 4 - 5 | 368 | 77 | |
BC003160.1 | - | 7 - 8 | 781 | 77 | |
BC093010.1 | - | 7 - 8 | 794 | 77 | |
X12548.1 | - | 7 - 8 | 790 | 77 |