Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
Chromosome Location
Chr.11: 13387996 - 13463297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297741.1 NP_001284670.1 3 - 4 689 143
NM_001297742.1 NP_001284671.1 3 - 4 879 143
NM_032320.6 NP_115696.2 4 - 5 847 143
XM_017018405.1 XP_016873894.1 3 - 4 1858 143
XM_017018406.1 XP_016873895.1 4 - 5 837 143
XM_017018407.1 XP_016873896.1 4 - 5 817 143
XM_017018408.1 XP_016873897.1 4 - 5 817 143
XM_017018409.1 XP_016873898.1 3 - 4 684 143
XM_017018410.1 XP_016873899.1 3 - 4 676 143
XM_017018411.1 XP_016873900.1 3 - 4 676 143
GenBank mRNA AF329102.2 - 4 - 5 847 143
AK292392.1 - 4 - 5 772 143
AK294259.1 - 3 - 4 668 143
AK294768.1 - 3 - 4 594 143
AK298226.1 - 3 - 4 564 143
AK310935.1 - 3 - 4 572 143
AK316163.1 - 3 - 4 879 143
AL832981.1 - 4 - 5 828 143
AY221959.2 - 4 - 5 890 143
BC005071.1 - 4 - 5 809 143

View Full Product Details