The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297741.1 | NP_001284670.1 | 7 - 8 | 1217 | 101 |
NM_001297742.1 | NP_001284671.1 | 7 - 8 | 1407 | 101 | |
NM_032320.6 | NP_115696.2 | 8 - 9 | 1375 | 101 | |
XM_017018405.1 | XP_016873894.1 | 8 - 9 | 2491 | 101 | |
XM_017018406.1 | XP_016873895.1 | 9 - 10 | 1470 | 101 | |
XM_017018407.1 | XP_016873896.1 | 9 - 10 | 1450 | 101 | |
XM_017018408.1 | XP_016873897.1 | 8 - 9 | 1345 | 101 | |
XM_017018409.1 | XP_016873898.1 | 8 - 9 | 1317 | 101 | |
XM_017018410.1 | XP_016873899.1 | 8 - 9 | 1309 | 101 | |
XM_017018411.1 | XP_016873900.1 | 7 - 8 | 1204 | 101 | |
GenBank mRNA | AF329102.2 | - | 8 - 9 | 1375 | 101 |
AK095946.1 | - | 7 - 8 | 1215 | 101 | |
AK292392.1 | - | 8 - 9 | 1300 | 101 | |
AK294259.1 | - | 7 - 8 | 1196 | 101 | |
AK298226.1 | - | 7 - 8 | 1092 | 101 | |
AK316163.1 | - | 7 - 8 | 1407 | 101 | |
AL832981.1 | - | 8 - 9 | 1356 | 101 | |
AY221959.2 | - | 8 - 9 | 1418 | 101 | |
BC005071.1 | - | 8 - 9 | 1337 | 101 | |
BC017072.1 | - | 3 - 4 | 352 | 101 | |
BC062633.1 | - | 4 - 5 | 485 | 101 |