The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170744.4 | NP_734465.2 | 7 - 8 | 1517 | 116 |
XM_011539453.1 | XP_011537755.1 | 8 - 9 | 1066 | 116 | |
XM_017015834.1 | XP_016871323.1 | 8 - 9 | 1076 | 116 | |
XM_017015835.1 | XP_016871324.1 | 7 - 8 | 1869 | 116 | |
GenBank mRNA | AB096256.1 | - | 7 - 8 | 1472 | 116 |
AK128132.1 | - | 7 - 8 | 1517 | 116 | |
AY358351.1 | - | 7 - 8 | 1463 | 116 |