The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003901.3 | NP_003892.2 | 4 - 5 | 485 | 122 |
XM_005270263.1 | XP_005270320.1 | 4 - 5 | 498 | 122 | |
XM_006718053.1 | XP_006718116.1 | 5 - 6 | 600 | 122 | |
XM_011540316.2 | XP_011538618.1 | 4 - 5 | 544 | 122 | |
XM_011540317.1 | XP_011538619.1 | 4 - 5 | 394 | 122 | |
XM_011540318.1 | XP_011538620.1 | 3 - 4 | 415 | 122 | |
GenBank mRNA | AB033078.1 | - | 4 - 5 | 463 | 122 |
AF144638.1 | - | 3 - 4 | 263 | 122 | |
AJ011304.2 | - | 4 - 5 | 440 | 122 | |
AK314615.1 | - | 4 - 5 | 394 | 122 | |
BC052991.1 | - | 4 - 5 | 473 | 122 |