Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COG7
Entrez Gene ID
91949
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
CDG2E
Chromosome Location
Chr.16: 23388493 - 23453215 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153603.3 NP_705831.1 4 - 5 798 81
XM_017023870.1 XP_016879359.1 4 - 5 841 81
GenBank mRNA AK093989.1 - 1 - 2 154 81
BC000549.2 - 4 - 5 753 81
BC037563.1 - 4 - 5 787 81
KU178837.1 - 4 - 5 601 81

View Full Product Details