The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303433.1 | NP_001290362.1 | 4 - 5 | 513 | 66 |
NM_003902.4 | NP_003893.2 | 3 - 4 | 450 | 66 | |
NR_130152.1 | 3 - 4 | 450 | 66 | ||
XM_011542391.2 | XP_011540693.1 | 4 - 5 | 384 | 66 | |
XM_011542392.2 | XP_011540694.1 | 4 - 5 | 418 | 66 | |
XM_011542393.2 | XP_011540695.1 | 3 - 4 | 355 | 66 | |
XM_017002739.1 | XP_016858228.1 | 5 - 6 | 406 | 66 | |
XM_017002740.1 | XP_016858229.1 | 4 - 5 | 384 | 66 | |
XM_017002741.1 | XP_016858230.1 | 4 - 5 | 402 | 66 | |
XM_017002742.1 | XP_016858231.1 | 4 - 5 | 418 | 66 | |
XM_017002743.1 | XP_016858232.1 | 3 - 4 | 359 | 66 | |
XM_017002744.1 | XP_016858233.1 | 3 - 4 | 359 | 66 | |
GenBank mRNA | AK300032.1 | - | 4 - 5 | 381 | 66 |
AK301581.1 | - | 3 - 4 | 318 | 66 | |
AK303568.1 | - | 3 - 4 | 317 | 66 | |
BC010083.2 | - | 3 - 4 | 285 | 66 | |
BC017247.2 | - | 3 - 4 | 314 | 66 | |
BQ649331.1 | - | 3 - 4 | 338 | 66 | |
U05040.2 | - | 3 - 4 | 275 | 66 |