The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303433.1 | NP_001290362.1 | 9 - 10 | 899 | 90 |
NM_003902.4 | NP_003893.2 | 8 - 9 | 836 | 90 | |
NR_130152.1 | 8 - 9 | 833 | 90 | ||
XM_011542391.2 | XP_011540693.1 | 9 - 10 | 770 | 90 | |
XM_011542392.2 | XP_011540694.1 | 9 - 10 | 801 | 90 | |
XM_011542393.2 | XP_011540695.1 | 8 - 9 | 741 | 90 | |
XM_017002739.1 | XP_016858228.1 | 10 - 11 | 792 | 90 | |
XM_017002740.1 | XP_016858229.1 | 9 - 10 | 770 | 90 | |
XM_017002741.1 | XP_016858230.1 | 9 - 10 | 785 | 90 | |
XM_017002742.1 | XP_016858231.1 | 9 - 10 | 801 | 90 | |
XM_017002743.1 | XP_016858232.1 | 8 - 9 | 742 | 90 | |
XM_017002744.1 | XP_016858233.1 | 8 - 9 | 742 | 90 | |
GenBank mRNA | AB209366.1 | - | 2 - 3 | 185 | 90 |
AK300032.1 | - | 9 - 10 | 767 | 90 | |
AK301581.1 | - | 8 - 9 | 701 | 90 | |
AK303568.1 | - | 8 - 9 | 700 | 90 | |
BC010083.2 | - | 8 - 9 | 671 | 90 | |
BC017247.2 | - | 8 - 9 | 697 | 90 | |
BQ649331.1 | - | 8 - 9 | 724 | 90 | |
U05040.2 | - | 8 - 9 | 661 | 90 |