The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242409.1 | NP_001229338.1 | 2 - 3 | 324 | 75 |
NM_022751.2 | NP_073588.1 | 2 - 3 | 324 | 75 | |
XM_017025919.1 | XP_016881408.1 | 2 - 3 | 358 | 75 | |
GenBank mRNA | AK025263.1 | - | 2 - 3 | 324 | 75 |
AK297430.1 | - | 1 - 2 | 119 | 75 | |
AL834491.1 | - | 2 - 3 | 272 | 75 | |
BC121067.1 | - | 2 - 3 | 291 | 75 | |
BC121068.1 | - | 2 - 3 | 291 | 75 |