The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271851.1 | NP_001258780.1 | 5 - 6 | 978 | 97 |
NM_022157.3 | NP_071440.1 | 5 - 6 | 1080 | 97 | |
GenBank mRNA | AF272035.1 | - | 5 - 6 | 908 | 97 |
AF323609.1 | - | 5 - 6 | 903 | 97 | |
AK023373.1 | - | 1 - 2 | 108 | 97 | |
AK298573.1 | - | 4 - 5 | 603 | 97 | |
AK298788.1 | - | 5 - 6 | 874 | 97 | |
AK315791.1 | - | 5 - 6 | 1005 | 97 | |
BC016668.1 | - | 5 - 6 | 925 | 97 | |
CA449149.1 | - | 2 - 3 | 97 |