The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286250.1 | NP_001273179.1 | 5 - 6 | 986 | 113 |
NM_001286251.1 | NP_001273180.1 | 4 - 5 | 887 | 113 | |
NM_001286252.1 | NP_001273181.1 | 6 - 7 | 1094 | 113 | |
NM_001323252.1 | NP_001310181.1 | 7 - 8 | 1141 | 113 | |
NM_001323253.1 | NP_001310182.1 | 6 - 7 | 1094 | 113 | |
NM_001323263.1 | NP_001310192.1 | 4 - 5 | 608 | 113 | |
NM_052904.3 | NP_443136.2 | 6 - 7 | 1094 | 113 | |
XM_005266813.4 | XP_005266870.1 | 4 - 5 | 499 | 113 | |
GenBank mRNA | AB067487.1 | - | 4 - 5 | 508 | 113 |
AK055292.1 | - | 6 - 7 | 980 | 113 | |
AK295479.1 | - | 5 - 6 | 811 | 113 | |
AK297240.1 | - | 4 - 5 | 631 | 113 | |
BC071627.1 | - | 6 - 7 | 969 | 113 |