The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031836.2 | NP_001027006.2 | 14 - 15 | 1608 | 119 |
NR_134267.1 | 14 - 15 | 1608 | 119 | ||
XM_011544424.1 | XP_011542726.1 | 14 - 15 | 1546 | 119 | |
XM_011544426.1 | XP_011542728.1 | 14 - 15 | 1546 | 119 | |
XM_011544427.1 | XP_011542729.1 | 14 - 15 | 1546 | 119 | |
XM_017013144.1 | XP_016868633.1 | 14 - 15 | 1546 | 119 | |
GenBank mRNA | AK302391.1 | - | 14 - 15 | 1559 | 119 |
BC028701.2 | - | 14 - 15 | 1549 | 119 |