The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022351.4 | NP_071746.1 | 1 - 2 | 290 | 117 |
XM_011517213.2 | XP_011515515.1 | 1 - 2 | 215 | 117 | |
GenBank mRNA | AF414126.2 | - | 2 - 3 | 379 | 117 |
AK095533.1 | - | 1 - 2 | 203 | 117 | |
AY224210.1 | - | 1 - 2 | 96 | 117 | |
BC068448.1 | - | 1 - 2 | 203 | 117 | |
DB481874.1 | - | 1 - 2 | 290 | 117 |