The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130783.4 | NP_570139.3 | 5 - 6 | 571 | 62 |
XM_005253217.3 | XP_005253274.1 | 7 - 8 | 755 | 62 | |
XM_006718372.3 | XP_006718435.1 | 7 - 8 | 842 | 62 | |
XM_006718373.3 | XP_006718436.1 | 6 - 7 | 828 | 62 | |
XM_011520459.2 | XP_011518761.1 | 6 - 7 | 718 | 62 | |
XM_017018530.1 | XP_016874019.1 | 6 - 7 | 593 | 62 | |
XM_017018531.1 | XP_016874020.1 | 5 - 6 | 515 | 62 | |
GenBank mRNA | AK027715.1 | - | 4 - 5 | 380 | 62 |
AK075547.1 | - | 3 - 4 | 304 | 62 | |
AY358087.1 | - | 4 - 5 | 494 | 62 | |
BC019342.2 | - | 3 - 4 | 343 | 62 |