Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETS1
Entrez Gene ID
2113
Gene Name
ETS proto-oncogene 1, transcription factor
Gene Aliases
ETS-1, EWSR2, c-ets-1, p54
Chromosome Location
Chr.11: 128458761 - 128587593 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143820.1 NP_001137292.1 4 - 5 421 118
NM_005238.3 NP_005229.1 2 - 3 520 118
XM_011542649.2 XP_011540951.1 5 - 6 557 118
XM_011542650.1 XP_011540952.1 3 - 4 1012 118
XM_011542651.1 XP_011540953.1 4 - 5 452 118
XM_011542652.1 XP_011540954.1 2 - 3 910 118
XM_017017314.1 XP_016872803.1 4 - 5 410 118
XM_017017315.1 XP_016872804.1 3 - 4 336 118
XM_017017316.1 XP_016872805.1 3 - 4 16527 118
XM_017017317.1 XP_016872806.1 4 - 5 456 118
GenBank mRNA AK291840.1 - 2 - 3 514 118
AK301405.1 - 2 - 3 519 118
BC017314.2 - 2 - 3 474 118
BP365215.1 - 2 - 3 518 118
BT019452.1 - 2 - 3 204 118
BT019453.1 - 2 - 3 204 118
BX640634.1 - 4 - 5 421 118
CR542254.1 - 2 - 3 204 118
J04101.1 - 2 - 3 323 118
X14798.1 - 2 - 3 482 118

View Full Product Details