Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETS1
Entrez Gene ID
2113
Gene Name
ETS proto-oncogene 1, transcription factor
Gene Aliases
ETS-1, EWSR2, c-ets-1, p54
Chromosome Location
Chr.11: 128458761 - 128587593 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143820.1 NP_001137292.1 8 - 9 1209 104
NM_001162422.1 NP_001155894.1 2 - 3 660 104
NM_005238.3 NP_005229.1 6 - 7 1308 104
XM_011542649.2 XP_011540951.1 9 - 10 1345 104
XM_011542650.1 XP_011540952.1 7 - 8 1800 104
XM_017017314.1 XP_016872803.1 8 - 9 1198 104
XM_017017315.1 XP_016872804.1 7 - 8 1124 104
XM_017017316.1 XP_016872805.1 7 - 8 17315 104
GenBank mRNA AK291840.1 - 6 - 7 1302 104
BT019452.1 - 6 - 7 992 104
BT019453.1 - 6 - 7 992 104
BX640634.1 - 8 - 9 1209 104
CR542254.1 - 6 - 7 992 104
J04101.1 - 6 - 7 1111 104
X14798.1 - 6 - 7 1270 104

View Full Product Details