Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROBO2
Entrez Gene ID
6092
Gene Name
roundabout guidance receptor 2
Gene Aliases
SAX3
Chromosome Location
Chr.3: 75906675 - 77649964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128929.3 NP_001122401.1 15 - 16 2554 152
NM_001290039.1 NP_001276968.1 15 - 16 2862 152
NM_001290040.1 NP_001276969.1 15 - 16 2862 152
NM_001290065.1 NP_001276994.1 15 - 16 2989 152
NM_002942.4 NP_002933.1 14 - 15 2850 152
XM_011533981.2 XP_011532283.1 15 - 16 2687 152
XM_011533982.1 XP_011532284.1 15 - 16 2687 152
XM_011533983.1 XP_011532285.1 15 - 16 2687 152
XM_011533984.1 XP_011532286.1 15 - 16 2687 152
XM_011533985.1 XP_011532287.1 15 - 16 2687 152
XM_017006986.1 XP_016862475.1 15 - 16 2910 152
XM_017006987.1 XP_016862476.1 15 - 16 2687 152
XM_017006988.1 XP_016862477.1 15 - 16 2687 152
XM_017006989.1 XP_016862478.1 14 - 15 2675 152
XM_017006990.1 XP_016862479.1 16 - 17 2566 152
XM_017006991.1 XP_016862480.1 15 - 16 2687 152
XM_017006992.1 XP_016862481.1 16 - 17 2566 152
XM_017006993.1 XP_016862482.1 14 - 15 2898 152
XM_017006994.1 XP_016862483.1 15 - 16 2687 152
XM_017006995.1 XP_016862484.1 15 - 16 2687 152
XM_017006996.1 XP_016862485.1 16 - 17 2566 152
XM_017006997.1 XP_016862486.1 16 - 17 2566 152
XM_017006998.1 XP_016862487.1 15 - 16 2910 152
XM_017006999.1 XP_016862488.1 15 - 16 2910 152
XM_017007000.1 XP_016862489.1 14 - 15 2898 152
XM_017007001.1 XP_016862490.1 15 - 16 2687 152
XM_017007002.1 XP_016862491.1 15 - 16 2687 152
XM_017007003.1 XP_016862492.1 14 - 15 2675 152
XM_017007004.1 XP_016862493.1 16 - 17 2566 152
XM_017007005.1 XP_016862494.1 16 - 17 2566 152
XM_017007006.1 XP_016862495.1 15 - 16 2910 152
GenBank mRNA AB046788.1 - 14 - 15 2213 152
AK074780.1 - 11 - 12 1500 152
AK301176.1 - 16 - 17 2439 152
BC144303.1 - 15 - 16 2275 152
BC146772.1 - 14 - 15 2213 152
DQ533873.1 - 15 - 16 2535 152
DQ533874.1 - 14 - 15 2850 152

View Full Product Details