Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROBO2
Entrez Gene ID
6092
Gene Name
roundabout guidance receptor 2
Gene Aliases
SAX3
Chromosome Location
Chr.3: 75906675 - 77649964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128929.3 NP_001122401.1 18 - 19 3027 83
NM_001290039.1 NP_001276968.1 18 - 19 3335 83
NM_001290040.1 NP_001276969.1 18 - 19 3335 83
NM_001290065.1 NP_001276994.1 18 - 19 3462 83
NM_002942.4 NP_002933.1 17 - 18 3323 83
XM_011533981.2 XP_011532283.1 18 - 19 3160 83
XM_011533982.1 XP_011532284.1 18 - 19 3160 83
XM_011533983.1 XP_011532285.1 18 - 19 3160 83
XM_011533984.1 XP_011532286.1 18 - 19 3160 83
XM_011533985.1 XP_011532287.1 18 - 19 3160 83
XM_017006986.1 XP_016862475.1 18 - 19 3383 83
XM_017006987.1 XP_016862476.1 18 - 19 3160 83
XM_017006988.1 XP_016862477.1 18 - 19 3160 83
XM_017006989.1 XP_016862478.1 17 - 18 3148 83
XM_017006990.1 XP_016862479.1 19 - 20 3039 83
XM_017006991.1 XP_016862480.1 18 - 19 3160 83
XM_017006992.1 XP_016862481.1 19 - 20 3039 83
XM_017006993.1 XP_016862482.1 17 - 18 3371 83
XM_017006994.1 XP_016862483.1 18 - 19 3160 83
XM_017006995.1 XP_016862484.1 18 - 19 3160 83
XM_017006996.1 XP_016862485.1 19 - 20 3039 83
XM_017006997.1 XP_016862486.1 19 - 20 3039 83
XM_017006998.1 XP_016862487.1 18 - 19 3383 83
XM_017006999.1 XP_016862488.1 18 - 19 3383 83
XM_017007000.1 XP_016862489.1 17 - 18 3371 83
XM_017007001.1 XP_016862490.1 18 - 19 3160 83
XM_017007002.1 XP_016862491.1 18 - 19 3160 83
XM_017007003.1 XP_016862492.1 17 - 18 3148 83
XM_017007004.1 XP_016862493.1 19 - 20 3039 83
XM_017007005.1 XP_016862494.1 19 - 20 3039 83
XM_017007006.1 XP_016862495.1 18 - 19 3383 83
GenBank mRNA AB046788.1 - 17 - 18 2686 83
AK301176.1 - 19 - 20 2912 83
BC064374.1 - 1 - 2 150 83
BC144303.1 - 18 - 19 2748 83
BC146772.1 - 17 - 18 2686 83
DQ533873.1 - 18 - 19 3008 83
DQ533874.1 - 17 - 18 3323 83

View Full Product Details