The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013353.2 | NP_037485.2 | 5 - 6 | 677 | 72 |
XM_011509449.1 | XP_011507751.1 | 5 - 6 | 559 | 72 | |
XM_017001089.1 | XP_016856578.1 | 4 - 5 | 1294 | 72 | |
XM_017001090.1 | XP_016856579.1 | 5 - 6 | 606 | 72 | |
GenBank mRNA | AF165217.1 | - | 5 - 6 | 554 | 72 |
AF177173.2 | - | 5 - 6 | 622 | 72 | |
AK092700.1 | - | 5 - 6 | 543 | 72 | |
AK313280.1 | - | 5 - 6 | 586 | 72 | |
BC017810.1 | - | 5 - 6 | 587 | 72 | |
BX647346.1 | - | 4 - 5 | 409 | 72 |