The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101417.3 | NP_001094887.1 | 6 - 7 | 878 | 98 |
NM_001101426.3 | NP_001094896.1 | 7 - 8 | 1028 | 98 | |
XM_006715770.3 | XP_006715833.1 | 7 - 8 | 1198 | 98 | |
XM_011515499.2 | XP_011513801.1 | 7 - 8 | 1173 | 98 | |
XM_011515500.2 | XP_011513802.1 | 6 - 7 | 1068 | 98 | |
XM_011515508.2 | XP_011513810.1 | 7 - 8 | 1530 | 98 | |
XM_011515509.2 | XP_011513811.1 | 7 - 8 | 2379 | 98 | |
XM_017012575.1 | XP_016868064.1 | 7 - 8 | 1173 | 98 | |
XM_017012577.1 | XP_016868066.1 | 6 - 7 | 1290 | 98 | |
XM_017012578.1 | XP_016868067.1 | 5 - 6 | 1705 | 98 | |
GenBank mRNA | DY654823.1 | - | 4 - 5 | 281 | 98 |