The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014436.2 | NP_001014436.1 | 3 - 4 | 349 | 65 |
NM_001122956.1 | NP_001116428.1 | 3 - 4 | 349 | 65 | |
NM_014063.6 | NP_054782.2 | 3 - 4 | 349 | 65 | |
XM_011515325.1 | XP_011513627.1 | 3 - 4 | 320 | 65 | |
GenBank mRNA | AF077353.1 | - | 3 - 4 | 289 | 65 |
AF197060.1 | - | 3 - 4 | 251 | 65 | |
AF218020.3 | - | 3 - 4 | 283 | 65 | |
AK027367.1 | - | 3 - 4 | 278 | 65 | |
AK293279.1 | - | 3 - 4 | 316 | 65 | |
AK302037.1 | - | 3 - 4 | 333 | 65 | |
BC011677.2 | - | 3 - 4 | 272 | 65 | |
BC031687.1 | - | 3 - 4 | 257 | 65 | |
CR457167.1 | - | 3 - 4 | 251 | 65 | |
DC404852.1 | - | 3 - 4 | 349 | 65 |