Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBNL
Entrez Gene ID
28988
Gene Name
drebrin like
Gene Aliases
ABP1, HIP-55, HIP55, SH3P7
Chromosome Location
Chr.7: 44044640 - 44061716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014436.2 NP_001014436.1 4 - 5 431 54
NM_001122956.1 NP_001116428.1 4 - 5 431 54
NM_001284313.1 NP_001271242.1 2 - 3 262 54
NM_014063.6 NP_054782.2 4 - 5 431 54
XM_011515325.1 XP_011513627.1 4 - 5 402 54
GenBank mRNA AB209486.1 - 2 - 3 118 54
AF077353.1 - 4 - 5 371 54
AF151364.1 - 4 - 5 343 54
AF197060.1 - 4 - 5 333 54
AF218020.3 - 4 - 5 365 54
AF250287.1 - 4 - 5 333 54
AK027367.1 - 4 - 5 360 54
AK293340.1 - 3 - 4 250 54
AK293698.1 - 2 - 3 227 54
AK296778.1 - 3 - 4 183 54
AK298102.1 - 3 - 4 183 54
AK302037.1 - 4 - 5 415 54
BC011677.2 - 4 - 5 354 54
BC031687.1 - 4 - 5 339 54
CR457167.1 - 4 - 5 333 54
DC404852.1 - 4 - 5 431 54

View Full Product Details