The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005886.2 | NP_005877.2 | 17 - 18 | 2015 | 58 |
XM_005255772.4 | XP_005255829.1 | 17 - 18 | 2096 | 58 | |
XM_006721121.3 | XP_006721184.1 | 17 - 18 | 2106 | 58 | |
XM_006721122.2 | XP_006721185.1 | 17 - 18 | 2230 | 58 | |
XM_006721123.3 | XP_006721186.1 | 17 - 18 | 2054 | 58 | |
XM_011522810.2 | XP_011521112.1 | 17 - 18 | 2744 | 58 | |
XM_017022860.1 | XP_016878349.1 | 17 - 18 | 2047 | 58 | |
XM_017022861.1 | XP_016878350.1 | 17 - 18 | 2221 | 58 | |
XM_017022862.1 | XP_016878351.1 | 17 - 18 | 2735 | 58 | |
XM_017022863.1 | XP_016878352.1 | 17 - 18 | 2184 | 58 | |
XM_017022864.1 | XP_016878353.1 | 17 - 18 | 2684 | 58 | |
GenBank mRNA | AB209250.1 | - | 14 - 15 | 2952 | 58 |
AF052432.1 | - | 16 - 17 | 1681 | 58 | |
BC001353.2 | - | 16 - 17 | 1664 | 58 | |
BT007022.1 | - | 16 - 17 | 1641 | 58 | |
CR456762.1 | - | 16 - 17 | 1641 | 58 |