Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
Chromosome Location
Chr.16: 57735717 - 57757250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005886.2 NP_005877.2 19 - 20 2208 59
XM_005255772.4 XP_005255829.1 19 - 20 2289 59
XM_006721121.3 XP_006721184.1 19 - 20 2299 59
XM_006721122.2 XP_006721185.1 19 - 20 2423 59
XM_006721123.3 XP_006721186.1 19 - 20 2247 59
XM_011522810.2 XP_011521112.1 19 - 20 2937 59
XM_017022860.1 XP_016878349.1 19 - 20 2240 59
XM_017022861.1 XP_016878350.1 19 - 20 2414 59
XM_017022862.1 XP_016878351.1 19 - 20 2928 59
XM_017022863.1 XP_016878352.1 19 - 20 2377 59
XM_017022864.1 XP_016878353.1 19 - 20 2877 59
GenBank mRNA AF052432.1 - 18 - 19 1874 59
BC001353.2 - 18 - 19 1857 59
BT007022.1 - 18 - 19 1834 59
CR456762.1 - 18 - 19 1834 59

View Full Product Details