The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005886.2 | NP_005877.2 | 8 - 9 | 1005 | 88 |
XM_005255772.4 | XP_005255829.1 | 8 - 9 | 1035 | 88 | |
XM_006721121.3 | XP_006721184.1 | 8 - 9 | 1036 | 88 | |
XM_006721122.2 | XP_006721185.1 | 8 - 9 | 1160 | 88 | |
XM_006721123.3 | XP_006721186.1 | 8 - 9 | 1035 | 88 | |
XM_011522810.2 | XP_011521112.1 | 8 - 9 | 1674 | 88 | |
XM_017022860.1 | XP_016878349.1 | 8 - 9 | 977 | 88 | |
XM_017022861.1 | XP_016878350.1 | 8 - 9 | 1160 | 88 | |
XM_017022862.1 | XP_016878351.1 | 8 - 9 | 1674 | 88 | |
XM_017022863.1 | XP_016878352.1 | 8 - 9 | 1174 | 88 | |
XM_017022864.1 | XP_016878353.1 | 8 - 9 | 1674 | 88 | |
GenBank mRNA | BC001353.2 | - | 7 - 8 | 654 | 88 |
BT007022.1 | - | 7 - 8 | 631 | 88 | |
CR456762.1 | - | 7 - 8 | 631 | 88 |