Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
Chromosome Location
Chr.1: 157676481 - 157700985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320333.1 NP_001307262.1 12 - 13 2262 76
NM_052939.3 NP_443171.2 12 - 13 2247 76
NR_135214.1 12 - 13 2280 76
NR_135215.1 12 - 13 2262 76
NR_135216.1 12 - 13 2167 76
NR_135217.1 11 - 12 1977 76
XM_006711145.1 XP_006711208.1 12 - 13 2177 76
GenBank mRNA AF416901.1 - 12 - 13 2126 76
AF416902.1 - 11 - 12 1841 76
AF416903.1 - 12 - 13 2144 76
AF416904.1 - 11 - 12 2360 76
AF416905.1 - 12 - 13 2031 76
AF459027.1 - 12 - 13 2247 76
AK098122.1 - 12 - 13 2177 76
AK125622.1 - 1 - 2 80 76
AL833391.1 - 6 - 7 2797 76
BC028933.1 - 12 - 13 2128 76

View Full Product Details