Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
Chromosome Location
Chr.1: 157676481 - 157700985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320333.1 NP_001307262.1 14 - 15 2364 79
NM_052939.3 NP_443171.2 14 - 15 2349 79
NR_135214.1 14 - 15 2382 79
NR_135215.1 14 - 15 2364 79
NR_135216.1 14 - 15 2269 79
NR_135217.1 13 - 14 2079 79
XM_006711145.1 XP_006711208.1 14 - 15 2279 79
GenBank mRNA AF416901.1 - 14 - 15 2228 79
AF416902.1 - 13 - 14 1943 79
AF416903.1 - 14 - 15 2246 79
AF416904.1 - 13 - 14 2462 79
AF416905.1 - 14 - 15 2133 79
AF459027.1 - 14 - 15 2349 79
AK301257.1 - 13 - 14 2485 79
AL833391.1 - 8 - 9 2899 79
AY043466.1 - 13 - 14 2153 79
BC028933.1 - 14 - 15 2230 79

View Full Product Details