Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IFT88
- Entrez Gene ID
-
8100
- Gene Name
- intraflagellar transport 88
- Gene Aliases
-
D13S1056E, DAF19, TG737, TTC10, hTg737
- Chromosome Location
-
Chr.13: 20566446 - 20691437 on Build GRCh38
View Full Product Details