Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
Chromosome Location
Chr.8: 25458569 - 25511308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317907.1 NP_001304836.1 2 - 3 609 69
NM_152562.3 NP_689775.2 2 - 3 609 69
XM_006716294.2 XP_006716357.1 2 - 3 609 69
XM_011544415.1 XP_011542717.1 2 - 3 609 69
XM_011544416.2 XP_011542718.1 2 - 3 303 69
XM_011544417.2 XP_011542719.1 2 - 3 360 69
XM_011544418.1 XP_011542720.1 2 - 3 588 69
GenBank mRNA AK299278.1 - 2 - 3 315 69
BC104450.1 - 2 - 3 123 69
BC104451.1 - 2 - 3 123 69
BM802460.1 - 2 - 3 278 69

View Full Product Details