The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127889.4 | NP_001121361.1 | 3 - 4 | 347 | 140 |
NM_016630.6 | NP_057714.1 | 3 - 4 | 528 | 140 | |
XM_005254437.4 | XP_005254494.1 | 4 - 5 | 561 | 140 | |
XM_017022297.1 | XP_016877786.1 | 3 - 4 | 896 | 140 | |
XM_017022298.1 | XP_016877787.1 | 4 - 5 | 986 | 140 | |
GenBank mRNA | AF208861.1 | - | 2 - 3 | 243 | 140 |
AF212231.1 | - | 3 - 4 | 315 | 140 | |
AK123725.1 | - | 1 - 2 | 760 | 140 | |
AK172849.1 | - | 3 - 4 | 360 | 140 | |
AK225527.1 | - | 3 - 4 | 360 | 140 | |
AK312943.1 | - | 3 - 4 | 362 | 140 | |
BC000244.1 | - | 3 - 4 | 398 | 140 | |
BI552731.1 | - | 3 - 4 | 498 | 140 | |
BP268459.1 | - | 3 - 4 | 521 | 140 | |
BP268875.1 | - | 3 - 4 | 140 | ||
BX446863.2 | - | 3 - 4 | 478 | 140 | |
DA680695.1 | - | 3 - 4 | 346 | 140 |