The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191022.1 | NP_001177951.1 | 8 - 9 | 1322 | 113 |
NM_006565.3 | NP_006556.1 | 10 - 11 | 2283 | 113 | |
XM_005255775.3 | XP_005255832.1 | 10 - 11 | 2283 | 113 | |
XM_017022868.1 | XP_016878357.1 | 10 - 11 | 2033 | 113 | |
XM_017022869.1 | XP_016878358.1 | 10 - 11 | 2032 | 113 | |
GenBank mRNA | AB209793.1 | - | 8 - 9 | 1156 | 113 |
AK093395.1 | - | 5 - 6 | 570 | 113 | |
AK314804.1 | - | 10 - 11 | 2281 | 113 | |
BC014267.2 | - | 10 - 11 | 2117 | 113 | |
BT009915.1 | - | 8 - 9 | 1839 | 113 | |
U25435.1 | - | 10 - 11 | 2130 | 113 |