The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006565.3 | NP_006556.1 | 4 - 5 | 1393 | 62 |
XM_005255775.3 | XP_005255832.1 | 4 - 5 | 1393 | 62 | |
XM_017022868.1 | XP_016878357.1 | 4 - 5 | 1143 | 62 | |
XM_017022869.1 | XP_016878358.1 | 4 - 5 | 1142 | 62 | |
GenBank mRNA | AK093395.1 | - | 2 - 3 | 196 | 62 |
BC014267.2 | - | 4 - 5 | 1227 | 62 | |
BT009915.1 | - | 2 - 3 | 949 | 62 | |
U25435.1 | - | 4 - 5 | 1240 | 62 |