The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206994.1 | NP_001193923.1 | 2 - 3 | 149 | 83 |
NM_001206995.1 | NP_001193924.1 | 2 - 3 | 253 | 83 | |
GenBank mRNA | AK296474.1 | - | 2 - 3 | 149 | 83 |
AK316003.1 | - | 3 - 4 | 263 | 83 | |
DC327075.1 | - | 2 - 3 | 258 | 83 |