The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014258.3 | NP_055073.2 | 23 - 24 | 2153 | 158 |
XM_011528487.2 | XP_011526789.1 | 23 - 24 | 2189 | 158 | |
XM_011528488.2 | XP_011526790.1 | 23 - 24 | 2141 | 158 | |
XM_011528489.2 | XP_011526791.1 | 23 - 24 | 2184 | 158 | |
XM_011528490.2 | XP_011526792.1 | 23 - 24 | 2165 | 158 | |
XM_011528493.2 | XP_011526795.1 | 22 - 23 | 2104 | 158 | |
XM_017027586.1 | XP_016883075.1 | 22 - 23 | 2385 | 158 | |
XM_017027587.1 | XP_016883076.1 | 22 - 23 | 2382 | 158 | |
XM_017027588.1 | XP_016883077.1 | 22 - 23 | 2386 | 158 | |
XM_017027589.1 | XP_016883078.1 | 22 - 23 | 2386 | 158 | |
XM_017027590.1 | XP_016883079.1 | 22 - 23 | 2078 | 158 | |
XM_017027591.1 | XP_016883080.1 | 14 - 15 | 1484 | 158 | |
XM_017027592.1 | XP_016883081.1 | 22 - 23 | 2387 | 158 | |
GenBank mRNA | AL080226.1 | - | 11 - 12 | 1087 | 158 |
BC040566.1 | - | 23 - 24 | 2189 | 158 | |
BC132864.1 | - | 22 - 23 | 2022 | 158 | |
BC132870.1 | - | 22 - 23 | 2024 | 158 | |
BC146798.1 | - | 23 - 24 | 2141 | 158 | |
BX648091.1 | - | 23 - 24 | 2107 | 158 | |
BX649108.1 | - | 23 - 24 | 2133 | 158 | |
Y08982.1 | - | 21 - 22 | 1932 | 158 |