The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014258.3 | NP_055073.2 | 25 - 26 | 2725 | 136 |
XM_011528487.2 | XP_011526789.1 | 25 - 26 | 2761 | 136 | |
XM_011528488.2 | XP_011526790.1 | 25 - 26 | 2713 | 136 | |
XM_011528489.2 | XP_011526791.1 | 25 - 26 | 2756 | 136 | |
XM_011528490.2 | XP_011526792.1 | 25 - 26 | 2737 | 136 | |
XM_011528493.2 | XP_011526795.1 | 24 - 25 | 2676 | 136 | |
XM_017027586.1 | XP_016883075.1 | 24 - 25 | 2957 | 136 | |
XM_017027587.1 | XP_016883076.1 | 24 - 25 | 2954 | 136 | |
XM_017027588.1 | XP_016883077.1 | 24 - 25 | 2958 | 136 | |
XM_017027589.1 | XP_016883078.1 | 24 - 25 | 2958 | 136 | |
XM_017027590.1 | XP_016883079.1 | 24 - 25 | 2650 | 136 | |
XM_017027591.1 | XP_016883080.1 | 16 - 17 | 2056 | 136 | |
GenBank mRNA | BC040566.1 | - | 25 - 26 | 2761 | 136 |
BC132864.1 | - | 24 - 25 | 2594 | 136 | |
BC132870.1 | - | 24 - 25 | 2596 | 136 | |
BX648091.1 | - | 25 - 26 | 2678 | 136 | |
BX649108.1 | - | 25 - 26 | 2704 | 136 | |
Y08982.1 | - | 23 - 24 | 2504 | 136 |