The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014258.3 | NP_055073.2 | 6 - 7 | 632 | 142 |
XM_011528487.2 | XP_011526789.1 | 6 - 7 | 668 | 142 | |
XM_011528488.2 | XP_011526790.1 | 6 - 7 | 620 | 142 | |
XM_011528489.2 | XP_011526791.1 | 6 - 7 | 663 | 142 | |
XM_011528490.2 | XP_011526792.1 | 6 - 7 | 644 | 142 | |
XM_017027586.1 | XP_016883075.1 | 5 - 6 | 864 | 142 | |
XM_017027587.1 | XP_016883076.1 | 5 - 6 | 861 | 142 | |
XM_017027588.1 | XP_016883077.1 | 5 - 6 | 865 | 142 | |
XM_017027589.1 | XP_016883078.1 | 5 - 6 | 865 | 142 | |
XM_017027592.1 | XP_016883081.1 | 5 - 6 | 866 | 142 | |
GenBank mRNA | AK308889.1 | - | 4 - 5 | 880 | 142 |
BC040566.1 | - | 6 - 7 | 668 | 142 | |
BC132864.1 | - | 5 - 6 | 501 | 142 | |
BC132870.1 | - | 5 - 6 | 503 | 142 | |
BC146798.1 | - | 6 - 7 | 620 | 142 | |
BX648091.1 | - | 6 - 7 | 586 | 142 | |
BX649108.1 | - | 6 - 7 | 612 | 142 | |
Y08982.1 | - | 4 - 5 | 411 | 142 |