Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYCP2
Entrez Gene ID
10388
Gene Name
synaptonemal complex protein 2
Gene Aliases
SCP-2, SCP2
Chromosome Location
Chr.20: 59863277 - 59933700 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014258.3 NP_055073.2 10 - 11 864 112
XM_011528487.2 XP_011526789.1 10 - 11 900 112
XM_011528488.2 XP_011526790.1 10 - 11 852 112
XM_011528489.2 XP_011526791.1 10 - 11 895 112
XM_011528490.2 XP_011526792.1 10 - 11 876 112
XM_011528493.2 XP_011526795.1 9 - 10 815 112
XM_017027586.1 XP_016883075.1 9 - 10 1096 112
XM_017027587.1 XP_016883076.1 9 - 10 1093 112
XM_017027588.1 XP_016883077.1 9 - 10 1097 112
XM_017027589.1 XP_016883078.1 9 - 10 1097 112
XM_017027590.1 XP_016883079.1 9 - 10 789 112
XM_017027591.1 XP_016883080.1 2 - 3 256 112
XM_017027592.1 XP_016883081.1 9 - 10 1098 112
GenBank mRNA AK308889.1 - 8 - 9 1112 112
BC040566.1 - 10 - 11 900 112
BC132864.1 - 9 - 10 733 112
BC132870.1 - 9 - 10 735 112
BC146798.1 - 10 - 11 852 112
BX648091.1 - 10 - 11 818 112
BX649108.1 - 10 - 11 844 112

View Full Product Details