The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030753.4 | NP_110380.1 | 3 - 4 | 750 | 76 |
GenBank mRNA | AB067628.1 | - | 3 - 4 | 601 | 76 |
AY009397.1 | - | 3 - 4 | 706 | 76 | |
BC112116.1 | - | 3 - 4 | 608 | 76 | |
BC112118.1 | - | 3 - 4 | 608 | 76 | |
BC114219.1 | - | 3 - 4 | 605 | 76 |