The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123066.3 | NP_001116538.2 | 11 - 12 | 2149 | 79 |
NM_001123067.3 | NP_001116539.1 | 7 - 8 | 1057 | 79 | |
NM_005910.5 | NP_005901.2 | 8 - 9 | 1144 | 79 | |
NM_016834.4 | NP_058518.1 | 6 - 7 | 970 | 79 | |
NM_016835.4 | NP_058519.3 | 10 - 11 | 2095 | 79 | |
XM_005257362.4 | XP_005257419.1 | 10 - 11 | 2187 | 79 | |
XM_005257364.4 | XP_005257421.1 | 9 - 10 | 2100 | 79 | |
XM_005257366.3 | XP_005257423.1 | 8 - 9 | 2055 | 79 | |
XM_005257367.4 | XP_005257424.1 | 9 - 10 | 1989 | 79 | |
XM_005257369.4 | XP_005257426.1 | 9 - 10 | 1122 | 79 | |
XM_005257370.4 | XP_005257427.1 | 8 - 9 | 1099 | 79 | |
XM_005257371.4 | XP_005257428.1 | 7 - 8 | 948 | 79 | |
GenBank mRNA | AY730549.1 | - | 6 - 7 | 735 | 79 |
BC114948.1 | - | 5 - 6 | 668 | 79 | |
X14474.1 | - | 5 - 6 | 678 | 79 |