The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123066.3 | NP_001116538.2 | 3 - 4 | 537 | 75 |
NM_001203252.1 | NP_001190181.1 | 3 - 4 | 537 | 75 | |
NM_005910.5 | NP_005901.2 | 3 - 4 | 537 | 75 | |
NM_016835.4 | NP_058519.3 | 3 - 4 | 537 | 75 | |
XM_005257362.4 | XP_005257419.1 | 3 - 4 | 317 | 75 | |
XM_005257365.4 | XP_005257422.1 | 3 - 4 | 317 | 75 | |
XM_005257367.4 | XP_005257424.1 | 3 - 4 | 317 | 75 | |
XM_005257368.4 | XP_005257425.1 | 3 - 4 | 317 | 75 | |
XM_005257369.4 | XP_005257426.1 | 3 - 4 | 317 | 75 |