The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177980.2 | NP_817089.1 | 11 - 12 | 1978 | 70 |
XM_011528969.2 | XP_011527271.1 | 11 - 12 | 1984 | 70 | |
XM_011528970.2 | XP_011527272.1 | 5 - 6 | 1000 | 70 | |
XM_017027994.1 | XP_016883483.1 | 11 - 12 | 2647 | 70 | |
XM_017027995.1 | XP_016883484.1 | 11 - 12 | 2648 | 70 | |
XM_017027996.1 | XP_016883485.1 | 11 - 12 | 2549 | 70 | |
XM_017027997.1 | XP_016883486.1 | 11 - 12 | 2648 | 70 | |
XM_017027998.1 | XP_016883487.1 | 11 - 12 | 2645 | 70 | |
XM_017027999.1 | XP_016883488.1 | 11 - 12 | 2645 | 70 | |
XM_017028000.1 | XP_016883489.1 | 10 - 11 | 1817 | 70 | |
XM_017028001.1 | XP_016883490.1 | 11 - 12 | 2645 | 70 | |
XM_017028002.1 | XP_016883491.1 | 11 - 12 | 2646 | 70 | |
GenBank mRNA | AF169690.1 | - | 3 - 4 | 442 | 70 |
AK055202.1 | - | 11 - 12 | 1967 | 70 | |
BC136306.1 | - | 11 - 12 | 1782 | 70 | |
BC143931.1 | - | 11 - 12 | 1782 | 70 |