The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005248299.3 | XP_005248356.1 | 5 - 6 | 790 | 148 |
GenBank mRNA | AK301220.1 | - | 5 - 6 | 789 | 148 |