The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281495.1 | NP_001268424.1 | 13 - 14 | 1574 | 94 |
NM_030877.4 | NP_110517.2 | 12 - 13 | 1473 | 94 | |
XM_011528917.2 | XP_011527219.1 | 10 - 11 | 1084 | 94 | |
GenBank mRNA | AF239607.1 | - | 12 - 13 | 1399 | 94 |
AF367471.1 | - | 9 - 10 | 990 | 94 | |
AF370431.1 | - | 12 - 13 | 1379 | 94 | |
AK074663.1 | - | 12 - 13 | 1386 | 94 | |
AK091216.1 | - | 12 - 13 | 2487 | 94 | |
AK130301.1 | - | 12 - 13 | 1386 | 94 | |
AK293420.1 | - | 13 - 14 | 1467 | 94 | |
BC022802.1 | - | 4 - 5 | 490 | 94 | |
BC036739.1 | - | 5 - 6 | 636 | 94 | |
BC121005.1 | - | 12 - 13 | 1394 | 94 | |
BC121006.1 | - | 12 - 13 | 1394 | 94 |